Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rftn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028713.1 | NP_082989.1 | 1 | 605 | ||
GenBank mRNA | AK012229.2 | - | 1 | 599 | ||
AK014318.1 | - | 1 | 357 | |||
AK031158.1 | - | 1 | 602 | |||
BC026871.1 | - | 1 | 169 | 3966594 | ||
BC038341.1 | - | 1 | 605 | 4218473 |