Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028770.2 | NP_083046.2 | 9 | 1381 | ||
GenBank mRNA | AK004811.2 | - | 8 | 1170 | ||
AK171114.1 | - | 1399 | ||||
BC119366.1 | - | 9 | 1381 | 8734115 | ||
BC120601.1 | - | 9 | 1381 | 40129524 |