Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933433C11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028961.1 | NP_083237.1 | 5 | 656 | ||
GenBank mRNA | AK017033.1 | - | 4 | 746 | ||
AK029738.1 | - | 5 | 614 | |||
BC050147.1 | - | 1 | 203 | |||
BC132286.1 | - | 5 | 656 | 40130563 | ||
BC138006.1 | - | 5 | 656 | 8861025 |