Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnks2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163635.1 | NP_001157107.1 | 27 | 4424 | ||
GenBank mRNA | AK017383.1 | - | 1 | 558 | ||
BC063101.1 | - | 10 | 1844 |