Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pyroxd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029011.2 | NP_083287.2 | 16 | 2528 | ||
GenBank mRNA | AK160239.1 | - | 16 | 2243 | ||
CB573340.1 | - | 1 | 520 |