Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntrk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270655.1 | NP_001257584.1 | 1, 2 | 292 | ||
NM_001270656.1 | NP_001257585.1 | 1, 2 | 292 | |||
NM_019248.2 | NP_062121.1 | 1, 2 | 292 | |||
XM_008759513.1 | XP_008757735.1 | 1, 2 | 283 | |||
GenBank mRNA | BC078844.1 | - | 1, 2 | 244 | 7121171 | |
L03813.1 | - | 1, 2 | 292 | |||
L14445.2 | - | 1, 2 | 241 | |||
L14446.1 | - | 1, 2 | 241 | |||
L14447.2 | - | 1, 2 | 241 | |||
S60953.1 | - | 1, 2 | 241 | |||
S62924.1 | - | 1, 2 | 241 | |||
S62933.1 | - | 1, 2 | 241 |