Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019277.2 | NP_062150.2 | 5, 6 | 504 | ||
XM_008760373.1 | XP_008758595.1 | 5, 6 | 558 | |||
XM_008760374.1 | XP_008758596.1 | 5, 6 | 2307 | |||
XM_008760375.1 | XP_008758597.1 | 5, 6 | 571 |