Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930568D16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029463.1 | NP_083739.1 | 4 | 260 | ||
GenBank mRNA | AK016248.1 | - | 4 | 261 |