Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177077.2 | NP_796051.2 | 22 | 2843 | ||
GenBank mRNA | AK220360.1 | - | 7 | 1117 | ||
BC030428.1 | - | 3 | 634 | |||
BC043037.1 | - | 5 | 934 | |||
BC059244.1 | - | 13 | 1754 | |||
BC144781.1 | - | 22 | 2786 | 9053329 | ||
BC145693.1 | - | 22 | 2786 | 40130996 |