Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spcs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029701.1 | NP_083977.1 | 5 | 2865 | ||
GenBank mRNA | AK028314.1 | - | 5 | 2827 | ||
AK037878.1 | - | 1 | 2260 | |||
AK048051.1 | - | 5 | 2852 | |||
BC019390.1 | - | 5 | 2798 | |||
BC054817.1 | - | 5 | 2830 | 5375997 |