Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kidins220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081378.1 | NP_001074847.1 | 32 | 6686 | ||
GenBank mRNA | AK122478.1 | - | 29 | 6279 | ||
AK151805.1 | - | 1 | 1369 | |||
BC030876.1 | - | 1 | 1862 | |||
BC044772.2 | - | 23 | 5805 | |||
BC046467.1 | - | 23 | 5805 | |||
BC059030.1 | - | 1 | 2114 | |||
BC094243.1 | - | 6 | 3182 | |||
KJ812113.1 | - | 9 | 3330 | |||
KJ812114.1 | - | 8 | 3297 | |||
KJ812115.1 | - | 7 | 3240 | |||
KJ812116.1 | - | 6 | 3207 | |||
KJ812117.1 | - | 6 | 3126 | |||
KJ812118.1 | - | 5 | 3093 |