Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033186.3 | NP_001028358.1 | 13 | 1787 | ||
NM_001177898.1 | NP_001171369.1 | 11 | 1630 | |||
NM_001177899.1 | NP_001171370.1 | 12 | 1739 | |||
GenBank mRNA | AK137942.1 | - | 13 | 1755 | ||
BC145402.1 | - | 12 | 1668 | 9053960 | ||
BC150764.1 | - | 12 | 1668 | 9087675 | ||
BC171961.1 | - | 12 | 1676 | 9054247 |