Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081679.1 | NP_001075148.1 | 1 | 29 | ||
GenBank mRNA | DQ854743.1 | - | 1 | 29 |