Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3galt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122993.1 | NP_001116465.1 | 4 | 5113 | ||
NM_033149.3 | NP_149161.1 | 3 | 4746 | |||
GenBank mRNA | AK004442.1 | - | 1 | 1236 | ||
BC057887.1 | - | 3 | 4744 | 4983830 |