Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYL1 SIRNA Assays ›
Gene Symbol:
Myl1
Gene Name:
myosin, light chain 1
Gene Aliases:
MLC3-f, MLCf, Mcl3, Mlc1, Mlc3
Chromosome Location:
Chr.9: 73937820 - 73958480 on Build Rnor_6.0
UniGene ID:
Rn.84920
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
51090
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Myl1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077656.1
NP_001071124.1
4
492
NM_020104.2
NP_064489.1
4
310
XM_008767294.1
XP_008765516.1
3
440
XM_008767295.1
XP_008765517.1
3
561
GenBank mRNA
CR755322.1
-
4
315
CR756356.1
-
4
310
FQ214608.1
-
4
497
FQ214631.1
-
4
479
FQ214638.1
-
4
587
FQ214656.1
-
4
328
FQ214675.1
-
4
586
FQ214712.1
-
4
500
FQ214723.1
-
4
500
FQ214727.1
-
4
500
FQ214742.1
-
4
500
FQ214939.1
-
4
497
FQ215002.1
-
4
586
FQ215050.1
-
4
500
FQ215083.1
-
4
497
FQ215086.1
-
4
330
FQ215201.1
-
4
497
FQ215229.1
-
4
591
FQ215325.1
-
4
586
FQ215340.1
-
4
328
FQ215418.1
-
4
586
FQ215435.1
-
4
497
FQ215492.1
-
4
479
FQ215500.1
-
3
1076
FQ215535.1
-
3
561
FQ215557.1
-
4
586
FQ215563.1
-
4
497
FQ215578.1
-
4
586
FQ215748.1
-
3
559
FQ215766.1
-
4
586
FQ215774.1
-
4
497
FQ215820.1
-
4
555
FQ215848.1
-
4
497
FQ215875.1
-
4
497
FQ215911.1
-
4
591
FQ215933.1
-
4
500
FQ216034.1
-
4
500
FQ216060.1
-
4
586
FQ216109.1
-
4
497
FQ216124.1
-
4
591
FQ216181.1
-
4
479
FQ216192.1
-
4
586
FQ216213.1
-
5
494
FQ216218.1
-
4
497
FQ216229.1
-
4
767
FQ216238.1
-
4
586
FQ216308.1
-
4
328
FQ216407.1
-
4
668
FQ216452.1
-
4
497
FQ216455.1
-
4
501
FQ216484.1
-
4
479
FQ216496.1
-
4
500
FQ216525.1
-
4
500
FQ216543.1
-
4
497
FQ216556.1
-
4
586
FQ216567.1
-
4
497
FQ216631.1
-
4
500
FQ216654.1
-
4
584
FQ216704.1
-
4
500
FQ216737.1
-
4
583
FQ216738.1
-
4
328
FQ217074.1
-
4
327
FQ217103.1
-
4
328
FQ217157.1
-
3
303
FQ217192.1
-
4
328
FQ217218.1
-
4
328
FQ217243.1
-
4
327
FQ217303.1
-
4
479
FQ217323.1
-
4
328
FQ217387.1
-
4
328
FQ217394.1
-
4
430
FQ217427.1
-
4
333
FQ217481.1
-
4
328
FQ217524.1
-
4
328
FQ217753.1
-
4
333
FQ217756.1
-
4
328
FQ217889.1
-
4
328
FQ217973.1
-
4
333
FQ217979.1
-
4
328
FQ218029.1
-
4
328
FQ218049.1
-
4
328
FQ223584.1
-
4
586
FQ223632.1
-
4
500
FQ223641.1
-
4
586
FQ223687.1
-
4
328
FQ223755.1
-
4
496
FQ223779.1
-
4
328
FQ223827.1
-
4
500
FQ223880.1
-
4
398
FQ223949.1
-
4
326
FQ223984.1
-
4
328
FQ224047.1
-
4
328
FQ224050.1
-
4
328
FQ224087.1
-
4
328
FQ224155.1
-
4
333
FQ224171.1
-
4
497
FQ224172.1
-
4
328
FQ224212.1
-
4
480
FQ224254.1
-
4
328
FQ224277.1
-
4
500
FQ224321.1
-
4
332
FQ224386.1
-
4
333
FQ224411.1
-
4
328
FQ224420.1
-
4
328
FQ224452.1
-
4
403
FQ224519.1
-
4
497
FQ224609.1
-
5
356
FQ224663.1
-
4
328
FQ224726.1
-
4
430
FQ224796.1
-
4
328
FQ224799.1
-
4
333
FQ224867.1
-
4
586
FQ224927.1
-
4
586
FQ224931.1
-
4
497
FQ224934.1
-
4
495
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cardiac muscle contraction
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-9ljwx:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0