Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU4F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004575.2 | NP_004566.2 | 2 | 1446 | ||
GenBank mRNA | AK314982.1 | - | 2 | 1449 | ||
BC136344.1 | - | 2 | 1353 | 9020331 | ||
BC136345.1 | - | 2 | 1353 | 9020332 | ||
EU439706.1 | - | 2 | 1182 | |||
U06233.1 | - | 2 | 1413 | |||
X71488.1 | - | 1 | 763 |