Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020543.4 | NP_065418.3 | 1 | 77 | ||
GenBank mRNA | AF218846.1 | - | 1 | 77 | ||
FJ694960.1 | - | 2 | 245 | |||
FJ694961.1 | - | 2 | 347 | |||
JN420349.1 | - | 3 | 442 | |||
JX494784.1 | - | 2 | 347 |
Set Membership: |
![]() |