Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021753.1 | NP_068521.1 | 2 | 198 | ||
GenBank mRNA | AJ251687.1 | - | 2 | 198 |
Gene Symbol: LOC100910889 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749058.3 | XP_003749106.1 | 2 | 857 |