Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022209.3 | NP_071545.2 | 2, 3 | 378 | ||
GenBank mRNA | AY251277.1 | - | 2, 3 | 330 | ||
BC078834.1 | - | 5, 6 | 587 | 7114294 | ||
D14421.1 | - | 2, 3 | 153 | |||
D38260.1 | - | 2, 3 | 173 | |||
FM080850.1 | - | 2, 3 | 378 |