Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022189.1 | NP_071525.1 | 2 | 166 | ||
GenBank mRNA | AF155044.1 | - | 2 | 166 |