Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022211.1 | NP_071547.1 | 2 | 444 | ||
GenBank mRNA | D64085.1 | - | 2 | 444 |