Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1rn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022194.2 | NP_071530.1 | 3, 4 | 333 | ||
XM_006233636.2 | XP_006233698.1 | 4, 5 | 2576 | |||
XM_006233637.2 | XP_006233699.1 | 4, 5 | 363 | |||
XM_006233638.2 | XP_006233700.1 | 3, 4 | 263 | |||
GenBank mRNA | BC070930.1 | - | 3, 4 | 327 | ||
CK364274.1 | - | 3, 4 | 357 |