Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081560.2 | NP_001075029.1 | 1 | 342 | ||
NM_001081562.2 | NP_001075031.1 | 1 | 342 | |||
NM_001288764.1 | NP_001275693.1 | 1 | 342 | |||
NM_001288766.1 | NP_001275695.1 | 1 | 342 | |||
NM_004409.4 | NP_004400.4 | 1 | 342 | |||
GenBank mRNA | AK302874.1 | - | 1 | 275 | ||
BC026328.1 | - | 1 | 337 | 4792104 | ||
BC062553.1 | - | 1 | 263 | 6480902 | ||
HY031977.1 | - | 1 | 285 | |||
L19268.1 | - | 1 | 264 | |||
M94203.1 | - | 1 | 15 | |||
S72883.1 | - | 1 | 173 |