Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022288.1 | NP_071624.1 | 429 | |||
XM_008766332.1 | XP_008764554.1 | 1 | 152 | |||
XM_008766333.1 | XP_008764555.1 | 1 | 152 | |||
XM_008766334.1 | XP_008764556.1 | 1 | 152 | |||
XM_008766335.1 | XP_008764557.1 | 1 | 152 | |||
GenBank mRNA | BC078831.1 | - | 431 | 7134461 | ||
U22296.1 | - | 429 |