Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164296.1 | NP_001157768.1 | 2, 3 | 275 | ||
NM_001164297.1 | NP_001157769.1 | 2, 3 | 210 | |||
NM_001164298.1 | NP_001157770.1 | 2, 3 | 269 | |||
NM_001164299.1 | NP_001157771.1 | 2, 3 | 301 | |||
NM_001164302.1 | NP_001157774.1 | 2, 3 | 255 | |||
NM_001164303.1 | NP_001157775.1 | 2, 3 | 208 | |||
NM_001164304.1 | NP_001157776.1 | 2, 3 | 468 | |||
NM_001164305.1 | NP_001157777.1 | 2, 3 | 185 | |||
NM_001164307.1 | NP_001157779.1 | 2, 3 | 282 | |||
NM_001164308.1 | NP_001157780.1 | 2, 3 | 342 | |||
NM_022401.2 | NP_071796.2 | 2, 3 | 661 | |||
XM_006241875.2 | XP_006241937.1 | 3, 4 | 634 | |||
GenBank mRNA | AY480022.1 | - | 2, 3 | 275 | ||
AY480023.1 | - | 2, 3 | 210 | |||
AY480024.1 | - | 2, 3 | 269 | |||
AY480025.1 | - | 2, 3 | 301 | |||
AY480026.1 | - | 2, 3 | 255 | |||
AY480027.1 | - | 2, 3 | 802 | |||
AY480028.1 | - | 2, 3 | 208 | |||
AY480029.1 | - | 2, 3 | 353 | |||
AY480030.1 | - | 2, 3 | 185 | |||
AY480031.1 | - | 2, 3 | 282 | |||
AY480032.1 | - | 2, 3 | 342 | |||
X59601.1 | - | 2, 3 | 611 |