Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022407.3 | NP_071852.2 | 4 | 409 | ||
GenBank mRNA | AF001896.1 | - | 4 | 409 | ||
AF001897.1 | - | 4 | 409 | |||
AF001898.1 | - | 4 | 409 | |||
BC061526.1 | - | 4 | 428 | 6921926 | ||
L42009.1 | - | 4 | 405 | |||
U79118.1 | - | 4 | 408 |