Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1cc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270983.1 | NP_001257912.1 | 5 | 856 | ||
NM_022498.2 | NP_071943.1 | 5 | 856 | |||
XM_008769212.1 | XP_008767434.1 | 6 | 743 | |||
GenBank mRNA | BC078825.1 | - | 5 | 783 | 7112668 | |
D90165.1 | - | 5 | 685 | |||
D90166.1 | - | 5 | 799 | |||
FM122849.1 | - | 2 | 168 |