Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhcr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022389.2 | NP_071784.1 | 7 | 1078 | ||
XM_006230892.2 | XP_006230954.1 | 6 | 826 | |||
XM_006230893.2 | XP_006230955.1 | 6 | 705 | |||
XM_006230894.2 | XP_006230956.1 | 7 | 725 | |||
XM_008760170.1 | XP_008758392.1 | 7 | 1812 | |||
XM_008760171.1 | XP_008758393.1 | 6 | 1473 | |||
XM_008760172.1 | XP_008758394.1 | 7 | 1372 | |||
XM_008760173.1 | XP_008758395.1 | 6 | 1352 | |||
GenBank mRNA | AB016800.1 | - | 5 | 769 | ||
AF071500.1 | - | 6 | 681 | |||
AF272393.1 | - | 7 | 1078 | |||
BC081688.1 | - | 5 | 769 | 7109221 |