Gene Symbol: IDI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317955.1 | NP_001304884.1 | 4, 5 | 760 | ||
NM_001317956.1 | NP_001304885.1 | 3, 4 | 894 | |||
NM_001317957.1 | NP_001304886.1 | 2, 3 | 721 | |||
NM_004508.3 | NP_004499.2 | 3, 4 | 777 | |||
NR_134300.1 | 2, 3 | 509 | ||||
NR_134301.1 | 2, 3 | 604 | ||||
XM_011519457.1 | XP_011517759.1 | 3, 4 | 1624 | |||
XM_017016191.1 | XP_016871680.1 | 2, 3 | 1451 | |||
GenBank mRNA | AF271720.1 | - | 2, 3 | 262 | ||
AK222875.1 | - | 3, 4 | 297 | |||
AK303669.1 | - | 3, 4 | 531 | |||
AK311110.1 | - | 2, 3 | 153 | |||
BC005247.2 | - | 2, 3 | 275 | |||
BC006999.2 | - | 2, 3 | 275 | |||
BC019227.2 | - | 2, 3 | 275 | |||
BC022418.2 | - | 2, 3 | 275 | |||
BC025375.2 | - | 2, 3 | 275 | |||
BC057827.1 | - | 4, 5 | 748 | 30337530 | ||
BC107893.1 | - | 3, 4 | 301 | |||
BE891119.1 | - | 3, 4 | 582 | |||
BT006761.1 | - | 2, 3 | 246 | |||
BX537663.1 | - | 2, 3 | 3321 | |||
BX648472.1 | - | 2, 3 | 1240 | |||
CD251816.1 | - | 3, 4 | 406 | |||
CN481934.1 | - | 2, 3 | 627 | |||
DA792902.1 | - | 2, 3 | 549 | |||
DA842794.1 | - | 3, 4 | 229 | |||
X17025.1 | - | 3, 4 | 296 |