Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU4F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004575.2 | NP_004566.2 | 2 | 1785 | ||
GenBank mRNA | BC136344.1 | - | 2 | 1692 | 9020331 | |
BC136345.1 | - | 2 | 1692 | 9020332 | ||
EU439706.1 | - | 2 | 1521 | |||
U06233.1 | - | 2 | 1752 |