Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022536.2 | NP_071981.1 | 2, 3 | 250 | ||
GenBank mRNA | AF071225.1 | - | 2, 3 | 250 | ||
BC061971.1 | - | 2, 3 | 279 | 5623342 | ||
FQ211358.1 | - | 2, 3 | 254 | |||
FQ212031.1 | - | 2, 3 | 279 | |||
FQ219792.1 | - | 2, 3 | 288 | |||
FQ219926.1 | - | 2, 3 | 300 | |||
FQ221020.1 | - | 2, 3 | 303 | |||
FQ221248.1 | - | 2, 3 | 298 | |||
FQ221362.1 | - | 2, 3 | 277 | |||
FQ222696.1 | - | 2, 3 | 412 | |||
FQ223162.1 | - | 2, 3 | 297 | |||
FQ228087.1 | - | 2, 3 | 293 | |||
FQ229174.1 | - | 2, 3 | 300 | |||
FQ229214.1 | - | 2, 3 | 294 | |||
FQ230004.1 | - | 304 | ||||
FQ231656.1 | - | 2, 3 | 293 | |||
FQ232077.1 | - | 2, 3 | 289 | |||
FQ232920.1 | - | 2, 3 | 303 |