Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251882.1 | NP_001238811.1 | 3 | 614 | ||
NM_001251885.1 | NP_001238814.1 | 2 | 191 | |||
NM_004624.3 | NP_004615.2 | 2 | 237 | |||
XM_005265437.2 | XP_005265494.1 | 2 | 206 | |||
XM_005265438.3 | XP_005265495.1 | 2 | 766 | |||
XM_011534079.1 | XP_011532381.1 | 3 | 965 | |||
XM_011534080.2 | XP_011532382.1 | 2 | 220 | |||
GenBank mRNA | AK056819.1 | - | 3 | 614 | ||
AK298115.1 | - | 2 | 160 | |||
AK314334.1 | - | 2 | 206 | |||
BC064424.1 | - | 2 | 193 | 5752796 | ||
EF577396.1 | - | 2 | 113 | |||
L13288.1 | - | 2 | 207 | |||
L20295.1 | - | 1 | 17 | |||
X75299.1 | - | 2 | 124 |
Set Membership: |
![]() |