Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022627.2 | NP_072149.1 | 5 | 571 | ||
XM_006233011.2 | XP_006233073.1 | 6 | 772 | |||
XM_006233012.2 | XP_006233074.1 | 6 | 672 | |||
GenBank mRNA | AF182717.1 | - | 5 | 573 |