Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gphn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022865.3 | NP_074056.2 | 2, 3 | 283 | ||
GenBank mRNA | EU735541.1 | - | 3, 4 | 198 | ||
X66366.2 | - | 2, 3 | 413 |