Gene Symbol: Inppl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270843.1 | NP_001257772.1 | 6 | 941 | ||
NM_022944.2 | NP_075233.1 | 7 | 997 | |||
GenBank mRNA | AB011439.1 | - | 6 | 941 | ||
AB025794.1 | - | 7 | 995 |