Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022678.1 | NP_073169.1 | 1 | 261 | ||
XM_006250328.2 | XP_006250390.1 | 3 | 1354 | |||
XM_006250330.2 | XP_006250392.1 | 2 | 1270 | |||
XM_006250331.2 | XP_006250393.1 | 2 | 749 | |||
XM_008769796.1 | XP_008768018.1 | 2 | 2001 | |||
XM_008769797.1 | XP_008768019.1 | 3 | 1133 | |||
XM_008769798.1 | XP_008768020.1 | 3 | 1353 | |||
XM_008769799.1 | XP_008768021.1 | 2 | 3272 | |||
XM_008769800.1 | XP_008768022.1 | 2 | 304 | |||
GenBank mRNA | AF221838.1 | - | 1 | 261 |