Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022846.2 | NP_074037.1 | 6 | 570 | ||
XM_006253906.1 | XP_006253968.1 | 6 | 630 | |||
GenBank mRNA | BC097316.1 | - | 6 | 571 | 7442457 | |
FQ219955.1 | - | 6 | 630 | |||
FQ220067.1 | - | 6 | 629 | |||
FQ220110.1 | - | 6 | 630 | |||
FQ222092.1 | - | 2 | 116 | |||
FQ228468.1 | - | 2 | 200 | |||
FQ229664.1 | - | 6 | 604 | |||
FQ229702.1 | - | 6 | 628 | |||
FQ229765.1 | - | 6 | 626 | |||
FQ229882.1 | - | 6 | 630 | |||
L06441.1 | - | 6 | 572 |