Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022861.1 | NP_074052.1 | 3708 | |||
GenBank mRNA | U24070.1 | - | 3708 |