Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gphn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022865.3 | NP_074056.2 | 5 | 560 | ||
GenBank mRNA | EU735541.1 | - | 6 | 475 | ||
X66366.2 | - | 5 | 690 |