Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BNIP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004331.2 | NP_004322.1 | 6 | 961 | ||
XM_005273617.3 | XP_005273674.1 | 6 | 865 | |||
GenBank mRNA | AB004788.1 | - | 6 | 882 | ||
AF060922.1 | - | 6 | 916 | |||
AF067396.1 | - | 6 | 919 | |||
AF079221.1 | - | 6 | 919 | |||
AF452712.2 | - | 6 | 934 | |||
AF536326.1 | - | 6 | 893 | |||
AK290213.1 | - | 6 | 877 | |||
AK300286.1 | - | 6 | 886 | |||
AK300727.1 | - | 6 | 754 | |||
AK307195.1 | - | 4 | 657 | |||
AK315870.1 | - | 6 | 838 | |||
AK316315.1 | - | 6 | 865 | |||
AL132665.1 | - | 6 | 935 | |||
BC001559.1 | - | 6 | 861 | 3459327 | ||
BC009603.1 | - | 6 | 929 | 4134765 |