Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbgt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023094.3 | NP_075582.3 | 2 | 119 | ||
GenBank mRNA | AB081649.1 | - | 3 | 194 | ||
AF264018.2 | - | 2 | 119 | |||
AF469945.1 | - | 2 | 119 |