Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024128.3 | NP_077042.2 | 5 | 1137 | ||
GenBank mRNA | BC060538.1 | - | 5 | 1140 | 6921928 |