Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmeff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023020.2 | NP_075409.1 | 5 | 789 | ||
GenBank mRNA | AJ250730.1 | - | 5 | 586 | ||
BC129093.1 | - | 5 | 789 | 8363015 |