Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Apaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023979.1 | NP_076469.1 | 2 | 745 | ||
GenBank mRNA | AF218388.1 | - | 2 | 200 | ||
AF320222.1 | - | 2 | 745 |