Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmur1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023100.1 | NP_075588.1 | 1, 2 | 851 | ||
GenBank mRNA | AB038649.1 | - | 1, 2 | 851 | ||
AF242873.1 | - | 1, 2 | 821 | |||
BC081796.1 | - | 878 |
Set Membership: |
![]() |