Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dctn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024130.1 | NP_077044.1 | 10 | 1303 | ||
GenBank mRNA | FQ223809.1 | - | 1023 | |||
X62160.1 | - | 10 | 1303 |