Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024156.2 | NP_077070.2 | 3 | 195 | ||
XM_006246340.2 | XP_006246402.1 | 3 | 933 | |||
XM_006246341.2 | XP_006246403.1 | 3 | 934 | |||
GenBank mRNA | BC072523.1 | - | 3 | 144 | 7105847 | |
CB585592.1 | - | 3 | 213 |