Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024357.3 | NP_077333.2 | 39 | 5160 | ||
XM_006251230.2 | XP_006251292.1 | 38 | 5098 | |||
XM_006251232.2 | XP_006251294.1 | 39 | 5146 | |||
XM_008770274.1 | XP_008768496.1 | 37 | 4612 | |||
GenBank mRNA | U18650.1 | - | 39 | 5004 |