Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024399.1 | NP_077375.1 | 2 | 416 | ||
XM_006246815.2 | XP_006246877.1 | 2 | 458 | |||
XM_006246817.2 | XP_006246879.1 | 2 | 458 | |||
XM_006246818.2 | XP_006246880.1 | 2 | 458 | |||
GenBank mRNA | AB023432.1 | - | 2 | 416 | ||
BC078813.1 | - | 2 | 418 | 7129010 | ||
BC078814.1 | - | 638 | 7133944 | |||
FQ232103.1 | - | 2 | 416 |