Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030826.3 | NP_110453.3 | 1 | 182 | ||
GenBank mRNA | BC058438.1 | - | 1 | 154 | 6918849 | |
CB579218.1 | - | 1 | 182 | |||
FQ211052.1 | - | 1 | 179 | |||
FQ227523.1 | - | 1 | 211 | |||
M21210.1 | - | 1 | 188 |